Search
Search

PRPF3| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046051.2 | NP_001039516.1 | 12 - 13 | 1723 | 111 | |
| GenBank mRNA | BC112709.1 | - | 12 - 13 | 1724 | 111 | |
Set Membership: |
|