Search
Search

MYDGF| Gene Symbol: MYDGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001164.1 | NP_001001164.1 | 2 - 3 | 235 | 84 | |
| GenBank mRNA | AY581200.1 | - | 2 - 3 | 235 | 84 | |
| BC102581.1 | - | 2 - 3 | 252 | 84 | ||
Set Membership: |
|