Search
Search

MYDGF| Gene Symbol: MYDGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001164.1 | NP_001001164.1 | 5 - 6 | 452 | 97 | |
| GenBank mRNA | AY581200.1 | - | 5 - 6 | 452 | 97 | |
| BC102581.1 | - | 5 - 6 | 469 | 97 | ||
Set Membership: |
|