Search
Search

TNFAIP6| Gene Symbol: TNFAIP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007813.2 | NP_001007814.1 | 3 - 4 | 457 | 124 | |
| GenBank mRNA | AJ854251.1 | - | 3 - 4 | 459 | 124 | |
| BC151789.1 | - | 3 - 4 | 471 | 124 | ||
Set Membership: |
|