Search
Search

SLC39A3| Gene Symbol: SLC39A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015521.1 | NP_001015521.1 | 4 - 5 | 825 | 60 | |
| GenBank mRNA | BC122570.1 | - | 3 - 4 | 626 | 60 | |
| BT020718.1 | - | 4 - 5 | 825 | 60 | ||
| BT021060.1 | - | 4 - 5 | 584 | 60 | ||
Set Membership: |
|