Search
Search

PARL| Gene Symbol: PARL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015596.1 | NP_001015596.1 | 4 - 5 | 504 | 138 | |
| GenBank mRNA | BC112869.1 | - | 4 - 5 | 512 | 138 | |
| BT020676.1 | - | 4 - 5 | 504 | 138 | ||
| BT026291.1 | - | 4 - 5 | 499 | 138 | ||
| BT029889.1 | - | 4 - 5 | 499 | 138 | ||
Set Membership: |
|