Search
Search

TNFSF13| Gene Symbol: TNFSF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034647.2 | NP_001029819.1 | 4 - 5 | 786 | 65 | |
| GenBank mRNA | BC103223.1 | - | 4 - 5 | 786 | 65 | |
| FJ539091.1 | - | 4 - 5 | 503 | 65 | ||
Set Membership: |
|