Search
Search

CD36| Gene Symbol: CD36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278621.1 | NP_001265550.1 | 5 - 6 | 692 | 101 | |
| NM_174010.3 | NP_776435.2 | 4 - 5 | 534 | 101 | ||
| GenBank mRNA | BC103112.1 | - | 5 - 6 | 681 | 101 | |
| D45364.1 | - | 2 - 3 | 276 | 101 | ||
| DV842657.1 | - | 4 - 5 | 696 | 101 | ||
| X91503.1 | - | 668 | 101 | |||
Set Membership: |
|