Search
Search

CSN3| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174294.2 | NP_776719.1 | 3 - 4 | 155 | 150 | |
| GenBank mRNA | BC102120.1 | - | 3 - 4 | 146 | 150 | |
| X00565.1 | - | 159 | 150 | |||
Set Membership: |
|