Search
Search

HSPE1| Gene Symbol: HSPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174346.2 | NP_776771.1 | 1 - 2 | 51 | 103 | |
| GenBank mRNA | BC102684.1 | - | 1 - 2 | 59 | 103 | |
| X69556.1 | - | 51 | 103 | |||
Set Membership: |
|