Search
Search

PLCB4| Gene Symbol: PLCB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166510.2 | NP_001159982.1 | 31 - 32 | 3214 | 58 | |
| GenBank mRNA | L13935.1 | - | 25 - 26 | 2722 | 58 | |
| L13936.1 | - | 26 - 27 | 2758 | 58 | ||
| L13937.1 | - | 24 - 25 | 2896 | 58 | ||
| L13938.1 | - | 25 - 26 | 2932 | 58 | ||
Set Membership: |
|