Search
Search

CRYBA4| Gene Symbol: CRYBA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174525.2 | NP_776950.1 | 3 - 4 | 235 | 73 | |
| GenBank mRNA | AY645777.1 | - | 3 - 4 | 198 | 73 | |
| M60328.1 | - | 3 - 4 | 235 | 73 | ||
Set Membership: |
|