Search
Search

GCSH| Gene Symbol: GCSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174844.1 | NP_777269.1 | 1 - 2 | 184 | 134 | |
| GenBank mRNA | BC120070.1 | - | 1 - 2 | 159 | 134 | |
| M58361.1 | - | 1 - 2 | 184 | 134 | ||
Set Membership: |
|