Search
Search
| Gene Symbol: MYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175050.1 | NP_778220.1 | 6 - 7 | 765 | 83 | |
| GenBank mRNA | D26147.1 | - | 6 - 7 | 765 | 83 | |
| D26148.1 | - | 6 - 7 | 765 | 83 | ||
| D26149.1 | - | 1 - 2 | 129 | 83 | ||
| M82978.1 | - | 1 - 2 | 158 | 83 | ||
Set Membership: |
|