Search
Search

FCGRT| Gene Symbol: FCGRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176657.1 | NP_788830.1 | 1 - 2 | 99 | 109 | |
| GenBank mRNA | AF139106.1 | - | 1 - 2 | 99 | 109 | |
| AF141017.1 | - | 1 - 2 | 99 | 109 | ||
Set Membership: |
|