Search
Search

CLCA3| Gene Symbol: CLCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181018.2 | NP_851361.1 | 5 - 6 | 617 | 147 | |
| GenBank mRNA | AF001261.1 | - | 5 - 6 | 617 | 147 | |
| AF001262.1 | - | 5 - 6 | 613 | 147 | ||
| AF001263.1 | - | 5 - 6 | 748 | 147 | ||
| AF001264.1 | - | 5 - 6 | 663 | 147 | ||
Set Membership: |
|