Search
Search

RPGRIP1| Gene Symbol: RPGRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181034.2 | NP_851377.1 | 7 - 8 | 785 | 61 | |
| GenBank mRNA | AF227258.1 | - | 7 - 8 | 785 | 61 | |
| AF265669.1 | - | 8 - 9 | 958 | 61 | ||
Set Membership: |
|