Search
Search

PRP6| Gene Symbol: PRP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205778.1 | NP_991347.1 | 4 - 5 | 594 | 132 | |
| GenBank mRNA | AB245482.1 | - | 4 - 5 | 594 | 132 | |
| BC149578.1 | - | 4 - 5 | 596 | 132 | ||
| X59504.1 | - | 594 | 132 | |||
Set Membership: |
|