Search
Search

AGO2| Gene Symbol: AGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205794.1 | NP_991363.1 | 11 - 12 | 1413 | 91 | |
| GenBank mRNA | AY533678.1 | - | 11 - 12 | 1413 | 91 | |
| BC151491.1 | - | 11 - 12 | 1521 | 91 | ||
Set Membership: |
|