Search
Search

NDUFA9| Gene Symbol: NDUFA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205817.1 | NP_991386.1 | 7 - 8 | 739 | 79 | |
| GenBank mRNA | BC102359.1 | - | 7 - 8 | 741 | 79 | |
| BT025401.1 | - | 7 - 8 | 693 | 79 | ||
| X59418.1 | - | 739 | 79 | |||
Set Membership: |
|