Search
Search

PSMC1| Gene Symbol: PSMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014855.1 | NP_001014855.1 | 9 - 10 | 1061 | 103 | |
| GenBank mRNA | BC123487.1 | - | 8 - 9 | 1042 | 103 | |
| BT020983.1 | - | 9 - 10 | 1061 | 103 | ||
Set Membership: |
|