Search
Search

PNPO| Gene Symbol: PNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014907.1 | NP_001014907.1 | 2 - 3 | 377 | 56 | |
| GenBank mRNA | BC103208.1 | - | 2 - 3 | 392 | 56 | |
| BT020917.1 | - | 2 - 3 | 377 | 56 | ||
Set Membership: |
|