Search
Search

CNPPD1| Gene Symbol: CNPPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015538.1 | NP_001015538.1 | 6 - 7 | 649 | 109 | |
| GenBank mRNA | BC113239.1 | - | 6 - 7 | 665 | 109 | |
| BT020928.1 | - | 6 - 7 | 649 | 109 | ||
Set Membership: |
|