Search
Search

SHMT1| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015553.1 | NP_001015553.1 | 10 - 11 | 1312 | 105 | |
| GenBank mRNA | BC112563.1 | - | 10 - 11 | 1298 | 105 | |
| BT020871.1 | - | 10 - 11 | 1312 | 105 | ||
Set Membership: |
|