Search
Search

SHMT1| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015553.1 | NP_001015553.1 | 11 - 12 | 1422 | 64 | |
| GenBank mRNA | BC112563.1 | - | 11 - 12 | 1408 | 64 | |
| BT020871.1 | - | 11 - 12 | 1422 | 64 | ||
Set Membership: |
|