Search
Search

TRIM10| Gene Symbol: TRIM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015607.1 | NP_001015607.1 | 4 - 5 | 937 | 90 | |
| GenBank mRNA | BC114866.1 | - | 4 - 5 | 956 | 90 | |
| BT020956.1 | - | 4 - 5 | 937 | 90 | ||
Set Membership: |
|