Search
Search

IFT22| Gene Symbol: IFT22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015612.1 | NP_001015612.1 | 4 - 5 | 453 | 89 | |
| GenBank mRNA | BC102614.1 | - | 3 - 4 | 368 | 89 | |
| BT020926.1 | - | 4 - 5 | 453 | 89 | ||
Set Membership: |
|