Search
Search

EIF3I| Gene Symbol: EIF3I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015628.1 | NP_001015628.1 | 6 - 7 | 566 | 66 | |
| GenBank mRNA | BC103177.1 | - | 6 - 7 | 581 | 66 | |
| BT021054.1 | - | 6 - 7 | 566 | 66 | ||
Set Membership: |
|