Search
Search

DTX2| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017945.1 | NP_001017945.1 | 7 - 8 | 1530 | 81 | |
| GenBank mRNA | BC103233.1 | - | 6 - 7 | 949 | 81 | |
| BT021146.1 | - | 7 - 8 | 1530 | 81 | ||
Set Membership: |
|