Search
Search

ALKBH2| Gene Symbol: ALKBH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024516.1 | NP_001019687.1 | 1 - 2 | 59 | 136 | |
| GenBank mRNA | BC120178.1 | - | 1 - 2 | 90 | 136 | |
| BT021573.1 | - | 1 - 2 | 59 | 136 | ||
Set Membership: |
|