Search
Search

IRF3| Gene Symbol: IRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029845.3 | NP_001025016.1 | 7 - 8 | 1310 | 80 | |
| GenBank mRNA | AJ879589.1 | - | 6 - 7 | 1104 | 80 | |
| BC102119.1 | - | 7 - 8 | 1311 | 80 | ||
| DQ103889.1 | - | 6 - 7 | 1090 | 80 | ||
| EU194377.1 | - | 6 - 7 | 1083 | 80 | ||
| KU641390.1 | - | 7 - 8 | 1151 | 80 | ||
Set Membership: |
|