Search
Search

PPARA| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034036.1 | NP_001029208.1 | 7 - 8 | 1386 | 84 | |
| GenBank mRNA | AF229356.1 | - | 5 - 6 | 1159 | 84 | |
| BT020756.1 | - | 7 - 8 | 1386 | 84 | ||
Set Membership: |
|