Search
Search

TRIM38| Gene Symbol: TRIM38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034574.1 | NP_001029746.1 | 3 - 4 | 625 | 67 | |
| GenBank mRNA | BC104589.1 | - | 3 - 4 | 647 | 67 | |
| BT021589.1 | - | 3 - 4 | 625 | 67 | ||
Set Membership: |
|