Search
Search

RPL23| Gene Symbol: RPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035014.2 | NP_001030186.1 | 3 - 4 | 254 | 72 | |
| GenBank mRNA | BC102556.1 | - | 3 - 4 | 255 | 72 | |
| BC148014.1 | - | 3 - 4 | 257 | 72 | ||
| DQ347609.1 | - | 1 - 2 | 77 | 72 | ||
Set Membership: |
|