Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRXL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040598.1 | NP_001035688.1 | 5 - 6 | 548 | 154 | |
GenBank mRNA | BC114900.1 | - | 5 - 6 | 467 | 154 | |
BT021811.1 | - | 5 - 6 | 548 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |