Search
Search

FABP5| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174315.3 | NP_776740.1 | 3 - 4 | 401 | 87 | |
| GenBank mRNA | BC105196.1 | - | 3 - 4 | 419 | 87 | |
| BT020981.1 | - | 3 - 4 | 401 | 87 | ||
| U55188.1 | - | 3 - 4 | 421 | 87 | ||
Set Membership: |
|