Search
Search

CSTF2| Gene Symbol: CSTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174685.2 | NP_777110.1 | 9 - 10 | 1059 | 78 | |
| GenBank mRNA | AY130297.1 | - | 9 - 10 | 1059 | 78 | |
| BC126543.1 | - | 10 - 11 | 1103 | 78 | ||
Set Membership: |
|