Search
Search

PFDN5| Gene Symbol: PFDN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174732.2 | NP_777157.1 | 4 - 5 | 299 | 123 | |
| GenBank mRNA | AF520958.1 | - | 4 - 5 | 299 | 123 | |
| BC102252.1 | - | 4 - 5 | 307 | 123 | ||
Set Membership: |
|