Search
Search

GGTA1| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177511.2 | NP_803477.1 | 5 - 6 | 583 | 78 | |
| GenBank mRNA | BC151691.1 | - | 4 - 5 | 295 | 78 | |
| J04989.1 | - | 5 - 6 | 583 | 78 | ||
Set Membership: |
|