Search
Search

RPS18| Gene Symbol: RPS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033614.2 | NP_001028786.1 | 4 - 5 | 323 | 106 | |
| GenBank mRNA | AB098889.1 | - | 4 - 5 | 304 | 106 | |
| AB098899.1 | - | 3 - 4 | 264 | 106 | ||
| AB099024.1 | - | 4 - 5 | 307 | 106 | ||
| BC102293.1 | - | 4 - 5 | 324 | 106 | ||
Set Membership: |
|