Search
Search

COL1A1| Gene Symbol: COL1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034039.2 | NP_001029211.1 | 26 - 27 | 1931 | 67 | |
| GenBank mRNA | BC105184.1 | - | 26 - 27 | 1932 | 67 | |
Set Membership: |
|