Search
Search

MRPL58| Gene Symbol: MRPL58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034493.1 | NP_001029665.1 | 2 - 3 | 245 | 104 | |
| GenBank mRNA | BC102162.1 | - | 2 - 3 | 245 | 104 | |
| BC134736.1 | - | 2 - 3 | 158 | 104 | ||
Set Membership: |
|