Search
Search

PINX1| Gene Symbol: PINX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034553.2 | NP_001029725.1 | 4 - 5 | 414 | 150 | |
| GenBank mRNA | BC102270.1 | - | 4 - 5 | 415 | 150 | |
| BT025471.1 | - | 4 - 5 | 416 | 150 | ||
Set Membership: |
|