Search
Search

SUCLG2| Gene Symbol: SUCLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034639.1 | NP_001029811.1 | 10 - 11 | 1186 | 88 | |
| GenBank mRNA | BC104559.1 | - | 10 - 11 | 1205 | 88 | |
Set Membership: |
|