Search
Search

SERPINC1| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034698.2 | NP_001029870.1 | 2 - 3 | 474 | 111 | |
| GenBank mRNA | BC102747.1 | - | 2 - 3 | 475 | 111 | |
Set Membership: |
|