Search
Search

PSMB8| Gene Symbol: PSMB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040480.2 | NP_001035570.1 | 3 - 4 | 428 | 152 | |
| GenBank mRNA | AF214525.1 | - | 2 - 3 | 180 | 152 | |
| BC102170.1 | - | 3 - 4 | 429 | 152 | ||
Set Membership: |
|