Search Thermo Fisher Scientific
Gene Symbol: RPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075581.2 | NP_001069049.1 | 1 | 166 | 81 | |
GenBank mRNA | BC102524.1 | - | 1 | 168 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |