Search
Search

MPZL1| Gene Symbol: MPZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076158.1 | NP_001069626.1 | 2 - 3 | 369 | 78 | |
| GenBank mRNA | BC108099.1 | - | 2 - 3 | 369 | 78 | |
| DQ375557.1 | - | 2 - 3 | 330 | 78 | ||
Set Membership: |
|