Search
Search

PLEKHO2| Gene Symbol: PLEKHO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076283.2 | NP_001069751.1 | 1 - 2 | 120 | 131 | |
| GenBank mRNA | BC109475.1 | - | 1 - 2 | 121 | 131 | |
Set Membership: |
|