Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076467.1 | NP_001069935.1 | 11 - 12 | 916 | 130 | |
NM_001370742.1 | NP_001357671.1 | 10 - 11 | 730 | 130 | ||
GenBank mRNA | BC102743.1 | - | 11 - 12 | 916 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |